missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Lithium perchlorate, anhydrous, 99.99% (metals basis)

Product Code. 11369738
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11369738 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11369738 Supplier Alfa Aesar™ Supplier No. 044226.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 400°C (decomposition)
Melting Point 236°C
CAS 3-9-7791
Color White
Physical Form Lump
Quantity 50 g
Molecular Formula ClH6LiO7
MDL Number MFCD00011079
UN Number UN1481
Merck Index 14,5539
Synonym lithium perchlorate, perchloric acid, lithium salt, lithiumperchlorate, unii-q86se98c9c, lithium 1+ ion perchlorate, lithium perchlorate, anhydrous, lithium cloricum, lithium per chlorate, liclo4, perchloric acid, lithium salt 1:1
InChI Key JAWYRNYHJJDXHX-UHFFFAOYSA-M
SMILES [Li+].[O-]Cl(=O)(=O)=O
IUPAC Name lithium;perchlorate
Molecular Weight (g/mol) 160.43
PubChem CID 23665649
Formula Weight 106.4
Percent Purity 99.99%
Packaging Ampule
Sensitivity Hygroscopic
Density 2.43g/mL
Assay (metals basis)
Chemical Name or Material Lithium perchlorate, Anhydrous
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.