missing translation for 'onlineSavingsMsg'
Learn More

Lithium Perchlorate, purum p.a, ≥98.0% (calc. based on dry substance, T), Honeywell™

Product Code. 15616060 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15616060 500 g Poly bottle 500g
15606060 100 g Poly bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15616060 Supplier Honeywell Research Chemicals Supplier No. 62580500G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum p.a.,≥98.0% (calc. based on dry substance, T), powder

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3-9-7791
Molecular Formula ClH6LiO7
Molecular Weight (g/mol) 160.43
MDL Number MFCD00011079
InChI Key JAWYRNYHJJDXHX-UHFFFAOYSA-M
Synonym lithium perchlorate, perchloric acid, lithium salt, lithiumperchlorate, unii-q86se98c9c, lithium 1+ ion perchlorate, lithium perchlorate, anhydrous, lithium cloricum, lithium per chlorate, liclo4, perchloric acid, lithium salt 1:1
PubChem CID 23665649
IUPAC Name lithium;perchlorate
SMILES [Li+].[O-]Cl(=O)(=O)=O

Specifications

Color White
pH 6.0 to 9.5 (at 104g/L 25°C)
Physical Form Granules
Quantity 500 g
Linear Formula LiClO4
UN Number UN1481
Formula Weight 106.39g/mol
Percent Purity ≥98.0% (calc. based on dry substance, T)
Packaging Poly bottle
Chemical Name or Material Lithium perchlorate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.