missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Lithium phthalocyanine

Product Code. 11368098
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11368098 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11368098 Supplier Alfa Aesar™ Supplier No. 039334.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 25510-41-2
Molecular Formula C32H18Li2N8
Molecular Weight (g/mol) 528.432
MDL Number MFCD00059094
InChI Key VEGOPYLKEARSAH-UHFFFAOYSA-N
Synonym dilithium phthalocyanine
PubChem CID 66736323
SMILES [Li].[Li].C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C(N4)N=C6C7=CC=CC=C7C(=N6)N=C8C9=CC=CC=C9C(=N8)N=C2N3

Specifications

Flash Point >110°C (230°F)
Quantity 5 g
Formula Weight 526.41
Physical Form Powder
Chemical Name or Material Lithium phthalocyanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.