missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Lithium pyruvate monohydrate

Product Code. 11427777
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11427777 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11427777 Supplier Alfa Aesar™ Supplier No. A16308.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 2922-61-4
Molecular Formula C3H3LiO3
Molecular Weight (g/mol) 93.994
MDL Number MFCD00150352
InChI Key OFJHGWPRBMPXCX-UHFFFAOYSA-M
Synonym lithium pyruvate, lithium 2-oxopropanoate, pyruvic acid lithium salt, propanoic acid, 2-oxo-, lithium salt, lithiumpyruvat, lithotab 2-oxopropanoate, propanoic acid, 2-oxo-, lithium salt 1:1, c3h3o3.li, lithium pyruvate h2o, lithium 1+ pyruvate
PubChem CID 23663618
IUPAC Name lithium;2-oxopropanoate
SMILES [Li+].CC(=O)C(=O)[O-]

Specifications

Quantity 10 g
Formula Weight 112.01 (94.00 Anhydrous)
Chemical Name or Material Lithium pyruvate monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.