missing translation for 'onlineSavingsMsg'
Learn More

Lithium stearate, 95%

Product Code. 10647941 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10647941

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4485-12-5 | C18H35LiO2 | 290.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4485-12-5
Molecular Formula C18H35LiO2
Molecular Weight (g/mol) 290.41
MDL Number MFCD00042032
InChI Key HGPXWXLYXNVULB-UHFFFAOYSA-M
Synonym lithium stearate, lithium octadecanoate, octadecanoic acid, lithium salt, stearic acid, lithium salt, lithalure, litholite, stavinor, lithium stearate, pure, stearic acid lithium salt, unii-p31mc94p70
PubChem CID 517357
IUPAC Name lithium;octadecanoate
SMILES [Li+].CCCCCCCCCCCCCCCCCC(=O)[O-]

Specifications

Melting Point 220°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula CH3(CH2)16COOLi
Quantity 25 g
Formula Weight 290.41
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Lithium stearate, 95%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.