missing translation for 'onlineSavingsMsg'
Learn More

Lithium Sulfate Monohydrate, ACS Reagent, ≥99% Dry Basis, Honeywell Fluka™

Product Code. 15615380 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15615380 500 g Poly Bottle 500g
15605380 100 g Poly Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15615380 Supplier Honeywell-Fluka Supplier No. 398152500G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ACS Reagent,≥99.0% dry basis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-25-7
Molecular Formula H2Li2O5S
Molecular Weight (g/mol) 127.95
MDL Number MFCD00149766
InChI Key RKGLUDFWIKNKMX-UHFFFAOYSA-L
Synonym lithium sulfate monohydrate, dilithium 1+ ion hydrate sulfate, dilithium sulfate hydrate, sufuric acid, dilithium slat, monohydrate, lithiumsulfatemonohydrate, dilithotab hydrate sulfate, lithium sulphate monohydrate, lithium sulfate, acs, ksc181s7n
PubChem CID 165815
IUPAC Name dilithium;sulfate;hydrate
SMILES [Li+].[Li+].O.[O-]S([O-])(=O)=O

Specifications

Color White
pH 5 to 7 (at 128g/L 25°C)
Quantity 500 g
Linear Formula Li2SO4 · H2O
UN Number NONH for all modes of transport
Solubility Information In water 128g/L at 20°C
Formula Weight 127.96g/mol
Percent Purity ≥99% (dry basis)
Packaging Poly Bottle
Appearance Crystalline
Density 2.06g/cm3
Chemical Name or Material Lithium sulfate monohydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.