missing translation for 'onlineSavingsMsg'
Learn More

Lithium Sulfate Monohydrate, (ACS Reagent Grade), MP Biomedicals

Product Code. 11422872 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11422872 500 g 500g
11412872 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11422872 Supplier MP Biomedicals Supplier No. 0215253990
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ACS Reagent grade chemical suitable for analytical and laboratory applications.

  • Grade: ACS reagent
  • Molecular Formula: H2Li2O5S
  • Molecular Weight: 127.96 g/mol
  • CAS Number: 10102-25-7
  • Melting Point: 130 °C
  • Density: 2.1 g/mL
  • Solubility: Soluble in cold water
  • Storage: Room temperature
  • Hazard Statement: H302

Chemical Identifiers

CAS 10102-25-7
Molecular Formula H2Li2O5S
Molecular Weight (g/mol) 127.95
MDL Number MFCD00149766
InChI Key RKGLUDFWIKNKMX-UHFFFAOYSA-L
Synonym lithium sulfate monohydrate, dilithium 1+ ion hydrate sulfate, dilithium sulfate hydrate, sufuric acid, dilithium slat, monohydrate, lithiumsulfatemonohydrate, dilithotab hydrate sulfate, lithium sulphate monohydrate, lithium sulfate, acs, ksc181s7n
PubChem CID 165815
IUPAC Name dilithium(1+) hydrate sulfate
SMILES [Li+].[Li+].O.[O-]S([O-])(=O)=O

Specifications

Quantity 500 g
Linear Formula Li2SO4·H2O
Solubility Information Soluble in water
ChEBI CHEBI:63300
Formula Weight 128
Grade Meets ACS reagent chemical requirements
Sensitivity Hygroscopic
Chemical Name or Material Lithium Sulfate Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.