missing translation for 'onlineSavingsMsg'
Learn More

Lithium Sulfate Monohydrate, Certified AR for Analysis, Fisher Chemical

Product Code. 10724571 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10724571 100 g Plastic powder jar 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10724571 Supplier Fisher Chemical Supplier No. L/2450/48
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 860°C
CAS 10102-25-7
Color White
Physical Form Powder/Solid
Quantity 100 g
Molecular Formula H2Li2O5S
MDL Number MFCD00149766
Synonym lithium sulfate monohydrate, dilithium 1+ ion hydrate sulfate, dilithium sulfate hydrate, sufuric acid, dilithium slat, monohydrate, lithiumsulfatemonohydrate, dilithotab hydrate sulfate, lithium sulphate monohydrate, lithium sulfate, acs, ksc181s7n
InChI Key RKGLUDFWIKNKMX-UHFFFAOYSA-L
SMILES [Li+].[Li+].O.[O-]S([O-])(=O)=O
IUPAC Name dilithium;sulfate;hydrate
Molecular Weight (g/mol) 127.95
PubChem CID 165815
Formula Weight 127.96g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • Lithium sulfate monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.