missing translation for 'onlineSavingsMsg'
Learn More

Lithium Sulfate Monohydrate, Honeywell™

Product Code. 15671240 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15671240 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15671240 Supplier Honeywell-Fluka Supplier No. 130291KG
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifica

CAS 10102-25-7
Quantity 1 kg
Molecular Formula H2Li2O5S
Linear Formula Li2SO4 · H2O
MDL Number MFCD00149766
UN Number NONH for all modes of transport
Synonym lithium sulfate monohydrate, dilithium 1+ ion hydrate sulfate, dilithium sulfate hydrate, sufuric acid, dilithium slat, monohydrate, lithiumsulfatemonohydrate, dilithotab hydrate sulfate, lithium sulphate monohydrate, lithium sulfate, acs, ksc181s7n
InChI Key RKGLUDFWIKNKMX-UHFFFAOYSA-L
SMILES [Li+].[Li+].O.[O-]S([O-])(=O)=O
IUPAC Name dilithium;sulfate;hydrate
Molecular Weight (g/mol) 127.95
PubChem CID 165815
Formula Weight 127.96g/mol
Percent Purity ≥98%
Chemical Name or Material Lithium Sulfate Monohydrate
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.