missing translation for 'onlineSavingsMsg'
Learn More

Lithium sulfide, 99.9% (metals basis)

Product Code. 11387567
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
250 g
Unit Size:
10g
250g
2g
50g
Product Code. Quantity unitSize
11387567 50 g 50g
11307577 2 g 2g
11397567 10 g 10g
11460310 250 g 250g
4 options
This item is not returnable. View return policy

Product Code. 11387567

Brand: Thermo Scientific Alfa Aesar 012839.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12136-58-2 | Li2S | 45.94 g/mol

It act as a superconductor and widely used in lithium-sulfur batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12136-58-2
Molecular Formula Li2S
Molecular Weight (g/mol) 45.94
MDL Number MFCD00011085
InChI Key GLNWILHOFOBOFD-UHFFFAOYSA-N
Synonym lithium sulfide, lithium sulfide li2s, lithium sulphide, anhydrous, li2s, zwdbutfcwlvlcq-uhfffaoysa-n, lithium sulfide trace metals basis
PubChem CID 64734
SMILES [Li+].[Li+].[S--]

Specifications

Melting Point 900°C to 975°C
Color Yellow
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.9% (Metal basis)
Solubility Information soluble in water and ethanol
IUPAC Name dilithium(1+) sulfanediide
Formula Weight 45.95
Odor Like rotten eggs
Sensitivity Moisture sensitive
Appearance Yellow powder
Density 1.66 g/cm3 at 20°C
Chemical Name or Material Lithium sulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.