missing translation for 'onlineSavingsMsg'
Learn More

Lithium tert-butoxide, 0.9M (8 wt.%) solution in 2-MeTHF, AcroSeal™

Product Code. 12635037 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12635037 100 mL 100mL
12615037 800 mL 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12635037 Supplier Thermo Scientific Acros Supplier No. 446841000
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1907-33-1 | C4H9LiO | 80.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1907-33-1 , 96-47-9
Molecular Formula C4H9LiO
Molecular Weight (g/mol) 80.05
InChI Key LZWQNOHZMQIFBX-UHFFFAOYSA-N
Synonym lithium tert-butoxide, lithium 2-methylpropan-2-olate, tert-butoxylithium, lithium tert-butylate, lithium tert-butanolate, tert-butyl alcohol, lithium salt, lithium t-butoxide, 2-methyl-2-propanol lithium salt, 2-propanol, 2-methyl-, lithium salt, lithium 1+ ion 2-methylpropan-2-olate
PubChem CID 23664764
IUPAC Name lithium;2-methylpropan-2-olate
SMILES [Li+].CC(C)(C)[O-]

Specifications

Quantity 100 mL
Linear Formula LiOCH(CH3)3
Flash Point -11°C
Formula Weight 80.05
Specific Gravity 0.86
Density 0.8600g/mL
Chemical Name or Material Lithium tert-butoxide
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lithium tert-butoxide, 8% in 2-MeTHF
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.