missing translation for 'onlineSavingsMsg'
Learn More

Lithium tetraborate, 99% (metals basis)

Product Code. 11376748
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
Product Code. Quantity unitSize
11376748 50 g 50g
11386748 250 g 250g
11366748 1 kg 1kg
3 options
This item is not returnable. View return policy

Product Code. 11376748

Brand: Thermo Scientific Alfa Aesar 013419.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12007-60-2 | B4Li2O7 | 169.11 g/mol

Used in the laboratory as LB (Lithium borate) buffer for gel electrophoresis of DNA and RNA. Employed in the borax fusion method to modify mineral powder samples for analysis by WDXRF (Wavelength Dispersive X-Ray Fluorescence) Spectroscopy. Acts as an ingredient in the manufacture of glasses and ceramics. Useful fod dissolving basic oxides such as CaO and MgO. A mixture of lithium metaborate and lithium tetraborate is useful for dissolving most of the inorganic oxides to enable spectrochemical analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12007-60-2
Molecular Formula B4Li2O7
Molecular Weight (g/mol) 169.11
MDL Number MFCD00011083
InChI Key RDZCISUGRQUANA-UHFFFAOYSA-N
IUPAC Name dilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
SMILES [Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2

Specifications

Melting Point 930°C
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 99% (Metals basis)
Solubility Information Moderately soluble in water.
Formula Weight 169.12
Density 2.4 g/cm3 at 20°C
Chemical Name or Material Lithium tetraborate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lithium tetraborate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H361d-Suspected of damaging the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Boron lithium oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.