missing translation for 'onlineSavingsMsg'
Learn More

Lithium Tetraborate, Flux Grade, BAKER INSTRA-ANALYZED™ Reagent, J.T. Baker™

Product Code. 15528304 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
Each
Product Code. Quantity Packaging unitSize
15528304 500 g Poly Bottle Each
1 options
This item is not returnable. View return policy

Product Code. 15528304

Brand: Avantor J.T.Baker 450301

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Acids and standards that are purified and characterized for use in trace element analysis in the low parts-per-billion (ppb) range

Specifications

CAS 12007-60-2
Quantity 500 g
Molecular Formula B4Li2O7
MDL Number MFCD00011083
InChI Key RDZCISUGRQUANA-UHFFFAOYSA-N
SMILES [Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
IUPAC Name dilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
Molecular Weight (g/mol) 169.11
Formula Weight 169.11
Grade BAKER INSTRA-ANALYZED™ Reagent
Packaging Poly Bottle
Specific Gravity 1L = 2.40kg
Chemical Name or Material Lithium Tetraborate, Flux Grade
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.