missing translation for 'onlineSavingsMsg'
Learn More

Lithium tetrachloropalladate(II)

Product Code. 11343337
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343337 5 g 5g
11353337 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11343337 Supplier Thermo Scientific Alfa Aesar Supplier No. 017316.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15525-45-8

It act as a catalyst in organic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15525-45-8
MDL Number MFCD00011082

Specifications

Physical Form Crystalline
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 262.09
Sensitivity Moisture sensitive
Chemical Name or Material Lithium tetrachloropalladate(II)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.