missing translation for 'onlineSavingsMsg'
Learn More

Lithium tetraphenylborate tris(1,2-dimethoxyethane) adduct

Product Code. 11393627
Change view
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.5g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11393627 2 g 2g
11383627 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393627 Supplier Thermo Scientific Alfa Aesar Supplier No. 035445.04
 Restricted Item
Estimated Shipment: 02-11-2026
Læg i indkøbskurv
Læg i indkøbskurv
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

CAS: 75965-35-4 | C36H50BLiO6 | 596.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 75965-35-4
Molecular Formula C36H50BLiO6
Molecular Weight (g/mol) 596.54
MDL Number MFCD00013311
InChI Key ADYUZFWVPWDPFK-UHFFFAOYSA-N
Synonym lithium tetraphenylborate tris 1,2-dimethoxyethane, tetraphenylboron lithium tris 1,2-dimethoxyethane, lithium tetraphenylborate tris 1,2-dimethoxyethane adduct, lithium tetraphenylborate 1--1,2-dimethoxyethane 1/1/3, lithium 1+ tris 1,2-dimethoxyethane tetraphenylborate
PubChem CID 23681136
IUPAC Name lithium;1,2-dimethoxyethane;tetraphenylboranuide
SMILES [Li+].[B-](C1=CC=CC=C1)(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.COCCOC.COCCOC.COCCOC

Tekniske data

Color White
Linear Formula (C6H5)4BLi·3CH3OCH2CH2OCH3
Quantity 2 g
UN Number UN1325
Sensitivity Moisture sensitive
Formula Weight 595.54 (326.18 Anhydrous)
Physical Form Powder
Chemical Name or Material Lithium tetraphenylborate tris(1,2-dimethoxyethane) adduct

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.