missing translation for 'onlineSavingsMsg'
Learn More

Lithium tri-sec-butylborohydride, 1.0M in THF, Thermo Scientific™

Product Code. 15401937
Change view
Click to view available options
Quantity:
25 mL
500 mL
Unit Size:
25mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15401937 25 mL 25mL
15421937 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15401937 Supplier Thermo Scientific Alfa Aesar Supplier No. 043845.AC
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

As a selective reducing agent

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38721-52-7
Molecular Formula C12H28BLi
Molecular Weight (g/mol) 190.11
MDL Number MFCD00011708
InChI Key ACJKNTZKEFMEAK-UHFFFAOYNA-N
Synonym lithium tri-sec-butylhydroborate, l-selectridesolution, lithium tri-sec-butylborohydride solution, l-selectride™ solution, lithium tri butan-2-yl boron 1-, lithium tri-sec-butylborohydride l-selectride , 1m in thf, lithium tri-sec-butylborohydride 1.0 m solution in thf, spcseal, lithium tri-sec-butylborohydride, 1.0m solution in thf, packaged under argon in resealable chemseal bottles
IUPAC Name lithium(1+) tris(butan-2-yl)boranuide
SMILES [Li+].CCC(C)[BH-](C(C)CC)C(C)CC

Specifications

Color Colorless to Yellow
Density 0.870g/mL
Flash Point −17°C (1°F)
Linear Formula LiB[CH(CH3)CH2CH3]3H
Quantity 25 mL
UN Number UN3394
Beilstein 4007327
Sensitivity Moisture Sensitive
Merck Index 14,8427
Formula Weight 190.11
Concentration or Composition (by Analyte or Components) 1.0M in THF
Physical Form Liquid
Chemical Name or Material Lithium tri-sec-butylborohydride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.