missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beskrivelse
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
Tekniske data
Tekniske data
| Melting Point | 319.0°C |
| Color | Gray-White to White |
| Density | 0.9 |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 93% min |
| Packaging | Glass Bottle packed in tin can |
| Linear Formula | LiAI[OC(CH3)3]3H |
| Quantity | 25 g |
| Fieser | 01,620; 02,251; 03,188; 04,312; 10,248; 12,286; 13,342; 16,197 |
| Solubility Information | Solubility in water: reacts violently. Other solubilities: 36g/100g thf (25°C),2g/100g diethyl ether (25°C),41g/100g diglyme (25°C) |
| Vis mere |
Sikkerhed og håndtering
Produktidentifikator
- Lithium tri-tert-butoxyaluminohydride
- Danger
- Flammable solid Category 1
- Skin corrosion/irritation Category 1 B
- Substance which, in contact with water, emits flammable gases Category 1
- H314-Causes severe skin burns and eye damage.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H228-Flammable solid.
- EUH014-Reacts violently with water.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?