missing translation for 'onlineSavingsMsg'
Learn More

Lithium tri-tert-butoxyaluminum hydride, nominally 1M in THF, packaged under Argon in resealable ChemSeal™ bottles

Product Code. 11339488
Change view
Click to view available options
Quantity:
10 mL
50 mL
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11339488 50 mL 50mL
11319488 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11339488 Supplier Thermo Scientific Alfa Aesar Supplier No. 043324.AD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 17476-04-9 | C12H28AlLiO3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17476-04-9
Molecular Formula C12H28AlLiO3
MDL Number MFCD00011532
Synonym Lithium hydridotris(tert-butoxy)aluminate

Specifications

Color Colorless
Density 0.904 g/mL
Flash Point −17°C (1°F)
Packaging ChemSeal™ bottles
Linear Formula Li[(CH3)3CO]3AlH
Quantity 50 mL
UN Number UN3399
Sensitivity Moisture sensitive
Solubility Information It is soluble in organic solvents at low temperatures
Formula Weight 254.28
Physical Form Liquid
Chemical Name or Material Lithium tri-tert-butoxyaluminum hydride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.