missing translation for 'onlineSavingsMsg'
Learn More

Lithium trifluoroacetate monohydrate, 97%

Product Code. 11369349
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11369349 10 g 10g
11359349 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11369349 Supplier Thermo Scientific Alfa Aesar Supplier No. 088938.09
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2923-17-3 | C2F3LiO2 | 119.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2923-17-3
Molecular Formula C2F3LiO2
Molecular Weight (g/mol) 119.96
MDL Number MFCD00013216,MFCD00149663
InChI Key HSFDLPWPRRSVSM-UHFFFAOYSA-M
Synonym lithium trifluoroacetate, acetic acid, trifluoro-, lithium salt, lithium 2,2,2-trifluoroacetate, lithiumtrifluoroacetate, acmc-1cfal, lithotab trifluoroacetate, acetic acid, 2,2,2-trifluoro-, lithium salt 1:1, lithium trifluoroacetate, monohydrate, lithium 1+ trifluoroacetate, lithio 2,2,2-trifluoroacetate
PubChem CID 23661853
IUPAC Name lithium;2,2,2-trifluoroacetate
SMILES [Li+].[O-]C(=O)C(F)(F)F

Specifications

Color White
Linear Formula LiCO2CF3·H2O
Quantity 10 g
UN Number UN2811
Sensitivity Hygroscopic
Formula Weight 137.97 (119.96 Anhydrous)
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Lithium trifluoroacetate monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.