missing translation for 'onlineSavingsMsg'
Learn More

Lithocholic acid, 95%

Product Code. 10571091 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10571091 5 g Glass bottle 5g
10511354 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10571091 Supplier Thermo Scientific Acros Supplier No. 229090050
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lithocholic acid, CAS # 434-13-9, a bacterial metabolite, is an oligo (ethylene glycol) and a secondary bile acid. | CAS: 434-13-9 | C24H40O3 | 376.581 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Lithocholic acid is a monohydroxy acid, and a secondary bile acid

Applications

  • Lithocholic acid can be used in studies that assess cholestasis
  • It can be used to study hepatic phospholipid regulation and homeostasis of bile acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 434-13-9
Molecular Formula C24H40O3
Molecular Weight (g/mol) 376.581
MDL Number MFCD00003682
InChI Key SMEROWZSTRWXGI-HVATVPOCSA-N
Synonym lithocholic acid, lithocolic acid, lithocholate, 3alpha-hydroxy-5beta-cholanic acid, 3alpha-hydroxy-5beta-cholan-24-oic acid, 3-hydroxycholan-24-oic acid, 3alpha-hydroxycholanic acid, 3-alpha-hydroxycholanic acid, 5beta-cholanic acid-3alpha-ol, 3alpha-hydroxy-5beta-cholanoic acid
PubChem CID 9903
ChEBI CHEBI:16325
IUPAC Name (4R)-4-[(3R,5R,8R,9S,10S,13R,14S,17R)-3-hydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
SMILES CC(CCC(=O)O)C1CCC2C1(CCC3C2CCC4C3(CCC(C4)O)C)C

Specifications

Melting Point 182°C to 188°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Beilstein 10, IV, 785
Merck Index 15, 5601
Specific Rotation Condition + 34.00 (20.00°C c=1.5,C2H5OH)
Specific Rotation On dry basis: +32° to +36° (20°C, 589nm) (c=1.5, C2H5OH)
Formula Weight 376.57
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material Lithocholic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.