missing translation for 'onlineSavingsMsg'
Learn More

Lithocholic acid, MedChemExpress

Product Code. 16492069
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492069 1 g 1g
16460798 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16492069 Supplier Medchem Express Supplier No. HYB0172
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Lithocholic acid is a toxic secondary bile acid, causes intrahepatic cholestasis, has tumor-promoting activity.

Lithocholic acid is a toxic secondary bile acid, causes intrahepatic cholestasis, has tumor-promoting activity.

Specifications

Chemical Name or Material Lithocholic acid
CAS 434-13-9
Color White
Purity Grade Notes Research
Molecular Formula C24H40O3
Quantity 1 g
Synonym 3α-Hydroxy-5β-cholanic acid
Solubility Information DMSO : 250 mg/mL (663.89 mM; Need ultrasonic) ∣Ethanol : 10 mg/mL (26.56 mM; ultrasonic and warming and heat to 60°C)
SMILES C[C@@]12[C@](CC[C@]2([H])[C@H](C)CCC(O)=O)([H])[C@@]3([H])[C@]([C@@]4([C@](C[C@H](O)CC4)([H])CC3)C)([H])CC1
Molecular Weight (g/mol) 376.57
Formula Weight 376.57
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.