missing translation for 'onlineSavingsMsg'
Learn More

Litmus, Indicator, pH 5-8, for titration, Honeywell Fluka™

Product Code. 15687910 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15687910 25 g 25g
15677910 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15687910 Supplier Honeywell-Fluka Supplier No. L738225G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Indicator, pH 5-8, for titration

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Litmus
Name Note Indicator, pH 5-8, for titration
CAS 1393-92-6, 64-17-5
CAS Max % ≤4.0000%, ≤100.0000%
CAS Min % ≥96.0000%
Molecular Formula I2
Linear Formula I2
MDL Number MFCD00011355
Grade Analytical Reagent
Packaging Glass Bottle
Quantity 25 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.