missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Litmus, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
- An acid-base indicator developed from various orcinol-containing lichen including Lecanora tartarea, Rocella fuciformis and Variolaria dealbata.
- The principal coloring matter in Litmus is Azolitmus, a substituted 7-hydroxy-2-phenoxazone derivative.
Specifications
Specifications
| CAS | 1393-92-6 |
| MDL Number | MFCD00131528 |
| Synonym | C.I. Natural Red 28, Lackmus, Tournesole, Chhadila |
| Solubility Information | Soluble in ethanol (3mg/mL); partly soluble water (10mg/mL-opaque dark blue solution) or alcohol. |
| Color | Blue |
| pH | 5 to 8 (Literature) |
| Quantity | 100 g |
| Physical Form | Powder |
| Chemical Name or Material | Litmus |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction