missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Litmus Paper Roll

Product Code. 15990872 Shop All Fisherbrand Products
Change view
Click to view available options
Type:
Litmus Paper, Blue
Litmus Paper, Red
Unit Size:
4 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
15990872 Litmus Paper, Red 4 units
15980872 Litmus Paper, Blue 4 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15990872
  • ONLINE EXCLUSIVE PRICE
    40.23 €
    20.66 € / 4 units
    Save 19.57 €
    48% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

Indicator papers without colour scale. For orientation and adjustment of pH values.

  • Impregnated indicator papers
  • Allows pH adjustment during special chemical procedures, not exact pH determination
  • No colour scale provided

Specifications

pH Range 5 to 8
Format Roll
For Use With (Application) For orientation and adjustment of pH values
Type Litmus Paper, Red
Length (Metric) 5 m
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.