missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Litmus, pure, indicator grade

Product Code. 10164160 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10164160 25 g Glass bottle 25g
10552924 100 g Glass bottle 100g
10494205 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10164160 Supplier Thermo Scientific Chemicals Supplier No. 190220250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 1393-92-6

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1393-92-6
MDL Number MFCD00131528

Specifications

Merck Index 15, 5603
Solubility Information Solubility in water: soluble. Other solubilities: partially soluble in alcohol
Infrared Spectrum Authentic
Grade Pure
Packaging Glass bottle
Visual Transition Interval from pH 4.8 (red) to pH 8.3 (blue)
Color Black/Blue to Blue
Quantity 25 g
Physical Form Powder or Granules
Chemical Name or Material Litmus

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.