missing translation for 'onlineSavingsMsg'
Learn More

Litmus Solution, Pure, Indicator Grade, Fisher Chemical™

Product Code. 11403633 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Unit Size:
2.5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403633 2.5 L 2.5 L
10284340 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11403633 Supplier Fisher Chemical Supplier No. L/2500L/17
Only null left
Add to basket
This item is not returnable. View return policy

Litmus Solution, Pure, Indicator Grade

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number 131528

Specifications

Packaging Amber glass bottle
Quantity 2.5 L
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.