missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LIVE/DEAD™ Fixable Near IR (876) Viability Kit, for 808 nm excitation
Description
- Fixable viability kits allow for clear determination of live and dead cells, enabling more precise analysis with both conventional and spectral flow cytometry
- Cells labeled with LIVE/DEAD Fixable viability dyes can be washed, fixed, permeabilized, and stained for intracellular antigens while maintaining distinct live and dead cell populations
- The narrow and unique emission spectra are ideal for expanding high-parameter flow cytometry experiments
- Designed to provide a bright signal with minimal overlap into adjacent channels, simplifying spectral compensation
- Since live and dead cells can be discriminated using a single flow cytometer channel, these viability dyes are an ideal choice for multi-color experiments.
- Their unique spectral fingerprints are a valuable addition to spectral flow cytometry experiments
- Dyes have been conveniently packaged in single-use vials to help ensure dye stability and performance over time
- Dissolve the dye in the included anhydrous DMSO and use immediately, or store at -20°C, avoiding freeze-thaw cycles
- The LIVE/DEAD Fixable Dead Cell Stain Sampler Kit provides one vial (40 assays) of each of eight available dead cell stains
Specifications
Specifications
| Quantity | 400 Assays |
| Description | LIVE/DEAD™ Fixable Near IR (876) Viability Kit, for 808 nm excitation |
| Format | Tubes |
| Color | Near-IR |
| Content And Storage | Contains 10 vials of LIVE/DEAD fixable dead cell stain and 2 vials of 500 μL DMSO. Store at -20°C. |
| Assay | Viability Assay |
| Compatible Cells | Eukaryotic Cells |
| Excitation/Emission | 849/876 nm |
| Flow Cytometer Laser Lines | 808 nm |
| Solubility | DMSO (Dimethylsulfoxide) |
| Show More |
Frequently Asked Questions (FAQs)
What is the concentration of the LIVE/DEAD Fixable dyes?
The concentration of LIVE/DEAD Fixable dyes is proprietary.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?