missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LiveBLAzer™ FRET-B/G Loading Kit with CCF4-AM

Product Code. 10798224
Change view
Click to view available options
Quantity:
1 mg
200 μg
5 mg
Unit Size:
1 set
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10798224 5 mg Each
10739934 200 μg 1 set
10563263 1 mg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10798224 Supplier Invitrogen™ Supplier No. K1030
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LiveBLAzer™ FRET-B/G Loading Kit with CCF4-AM

Background:
GeneBLAzer™ cell-based assays utilize the membrane-permeant ester forms (CCF2-AM and CCF4-AM) of the negatively charged fluorescent beta-lactamase substrates, CCF2 and CCF4. These lipophilic esters readily enter the cell, where cleavage by endogeneous cytoplasmic esterases rapidly converts them into their negatively charged forms, thereby trapping them in the cytosol.

Detection of GeneBLAzer™ assays is FRET-based. Each substrate is labeled with two fluorophores that form an efficient FRET pair. In the absence of beta-lactamase activity, exciting the coumarin at 409 nm in the intact CCF2 molecule results in FRET to the fluorescein, which emits a green fluorescence signal at 518 nm (Figure 1). In the presence of beta-lactamase activity, however, cleavage of CCF2 spatially separates the two dyes and disrupts FRET, so that exciting the coumarin at 409 nm now produces a blue fluorescence signal at 447 nm. This blue signal can be readily observed under a microscope and can also be detected as an increase in the blue channel readout on fluorescent microplate readers.

The CCF2-AM and CCF4-AM substrates are essential assay components for the GeneBLAzer™ platform. These substrates are fully compatible with flow cytometry, speeding the time to clone selection. Ratiometric analysis of the blue and green signals reduces well-to-well variation due to differences in cell numbers and substrate loading, leading to high Z´-factor values and low coefficients of variation (CVs).

CCF2-AM and CCF4-AM differ by two carbons in the bridge linking the coumarin moiety to the lactam ring. Both are in the membrane-permeable, esterified forms, and can be used for assays in intact cells. CCF4-AM has better solubility properties (soluble for >24 hours) than CCF2-AM and is thus best suited for screening applications. In addition, CCF4-AM has slightly better FRET and thus slightly lower background than CCF2-AM.

CCF2-FA is essentially the CCF2 substrate without the esters found in the AM version. CCF2-FA is de-esterified and used in cell lysate applications, bypassing loading across the cell membrane and de-esterification steps. Cell lysates are the preferred method for applications using cells that contain a cell wall. CCF2-FA can also be used as a control to acquire the excitation and emission spectra for CCF2-AM and CCF4-AM.
TRUSTED_SUSTAINABILITY

Specifications

Cell Permeability Cell-Permeant
Content And Storage GeneBLAzer™ beta-lactamase substrates are supplied as loading kits (Table 1) and are available in bulk sizes. Loading kits include CCF2-AM or CCF4-AM, DMSO (for Solution A), Solution B, and Solution C. CCF2-AM and CCF4-AM are provided dry, and should be stored at -20°C, desiccated and protected from light. DMSO (for Solution A), Solution B, and Solution C should be stored at 22-25°C, protected from direct light. Slightly cooler temperatures may cause a white precipitate to form in Solution B.
Detection Method Fluorescence
For Use With (Equipment) Flow Cytometer, Microplate Reader
Product Line LiveBLAzer
Quantity 5 mg
Substrate Properties Chemical Substrate
Substrate Type Beta-Lactamase Substrate
Substrate CCF4-AM
Type FRET-B/G Loading Kit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.