missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ LJI308
GREENER_CHOICE

Product Code. 18787156
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18787156

Brand: Tocris Bioscience™ 6754/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent pan-ribosomal S6 protein kinase (RSK) inhibitor (IC50 values are 4 to 13 nM)

LJI308 is a potent pan-ribosomal S6 protein kinase (RSK) inhibitor (IC50 values are 4 to 13 nM). Displays selectivity for RSK isoforms over other kinases. Inhibits RSK-dependent Y-box binding protein 1 (YB-1) phosphorylation in vitro. Suppresses growth of triple negative breast cancer cell lines.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,6-Difluoro-4-[4-[4-(4-morpholinyl)phenyl]-3-pyridinyl]phenol
CAS 1627709-94-7
Quantity 25 mg
Molecular Formula C21H18F2N2O2
Formula Weight 368.38
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.