Learn More
Heinz Herenz LMS-Tube LIM Broth
Product Code. 30158380
Change view
Click to view available options
Sterility:
Sterile
Unit Size:
Pack of 100
Product Code. Sterility Unit Size
30158380 Sterile Pack of 100
1 options
Product Code. 30158380 Supplier Heinz Herenz Supplier No. 1020316
  • ONLINE EXCLUSIVE PRICE
    44.56 €
    27.72 € / Pack of 100
    Save 16.84 €
    37% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Without swab. Includes blue screw cap and contains 2 mL of liquid LIM Broth medium. Sterile.
Our transport and collection swabs contain special applicators and test tubes with different liquid media, customized for the transportation and preservation of microbiological samples.
  • The liquid medium allows the division of the sample material and thus several tests from one sample.
  • The ready to use devices are supplied in a peelpack which is easy to open.
  • The breakpoint in the shaft of the applicator enables a safe and hygienic workflow.
  • The standard size of the test tubes with screwcap simplified the use with automatic analyzers.
  • There are labels on the test tubes for a accurate sample identification.
  • The items are produced according to the conditions of GMP (Good Manufacturing Practice) and certified according to DIN EN ISO 13485. This ensures a high quality.
  • Our swabs are sterile (SAL 106), pyrogen free and noncytotoxic.
Specifications
Product Type Transport and Collection Swab
Format Swab
Sterility Sterile
Color White