missing translation for 'onlineSavingsMsg'
Learn More

Loading Solutions

Product Code. 10475263
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10475263 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10475263 Supplier Invitrogen Supplier No. K1026
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Loading Solutions

This kit contains the solutions needed to conveniently load the FRET-based, beta-lactamase substrates, CCF2-AM and CCF4-AM, into cells.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Product Line GeneBLAzer
Quantity 1 Kit
Shipping Condition Room Temperature
Type Loading Solution

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.