Learn More
Brady™ Lockout Starter Kit
Product Code. 15653547 Shop All Brady Products
Change view
Click to view available options
Color:
Red/Silver
Unit Size:
Each
Product Code. Color Unit Size
15653547 Red/Silver Each
1 options
Product Code. 15653547 Supplier Brady™ Supplier No. 806183
  • ONLINE EXCLUSIVE PRICE
    543.00 €
    365.65 € / Pack of 1
    Save 177.35 €
    32% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The combination lockout kit gives you everything you need to lock out the most common mechanical and electrical control points.
  • Kit for valve and electrical lockout
Specifications
Alternate Part Number Lockout Starter Kit (NL)
Color Red/Silver
Includes 1 x 050899 - Large Universal Valve Lockout Base Clamping Unit, 1 x 065403 - Large Blocking Arm, 1 x 051395 - Cable attachment with 2.44 m sheathed cable - Cable dia. 3.25 mm, 1 x 064057 - Adjustable Gate Valve Lockout, 1 x 050943 - All purpose Cable Lockout with Vinyl Coated Metal Cable - 2.44 m, 065675 - Lockout Devices for Plug Connections - Large, 1 x 065396 - No Hole Circuit Breaker Lockout - 277 V Device, 1 x 065397 - No Hole Circuit Breaker - 480 - 600 V Device, 1 x 090891 480 - 600 V Breaker Lockout kit, 1 x 805853 Miniature Breaker Lockout Demo Kit, 1 x 236916 - Nylon Lockout Hasp, - 6 x 051339 - Safety Padlock - Standard, 12 x 805840 - Safety Lockout, 1 x 099162 Lockout Duffel Bag, 10 x 2562395 - DO NOT OPERATE - Maintenance Department Tag (DE), 1 x 087052 - Permanent Marker - fine