missing translation for 'onlineSavingsMsg'
Learn More

Lofexidine Hydrochloride, TRC

Product Code. 30481322
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481322 50 mg 50mg
30479654 5 mg 5mg
30474939 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30481322 Supplier Toronto Research Chemicals Supplier No. TRCL46939050MG
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21498-08-8
Molecular Formula C11 H12 Cl2 N2 O . Cl H
Molecular Weight (g/mol) 295.59
Synonym 1H-Imidazole, 2-[1-(2,6-dichlorophenoxy)ethyl]-4,5-dihydro-, hydrochloride (1:1), 1H-Imidazole, 2-[1-(2,6-dichlorophenoxy)ethyl]-4,5-dihydro-, monohydrochloride (9CI), 2-Imidazoline, 2-[1-(2,6-dichlorophenoxy)ethyl]-, monohydrochloride (8CI), Baq 168, Britlofex, Lofetensin, Lofexidine hydrochloride, Loxacor, MDL 14042
IUPAC Name 2-[1-(2,6-dichlorophenoxy)ethyl]-4,5-dihydro-1H-imidazole;hydrochloride
SMILES Cl.CC(Oc1c(Cl)cccc1Cl)C2=NCCN2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C11H12Cl2N2O.ClH/c1-7(11-14-5-6-15-11)16-10-8(12)3-2-4-9(10)13;/h2-4,7H,5-6H2,1H3,(H,14,15);1H
Formula Weight 294.0093
Chemical Name or Material Lofexidine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.