missing translation for 'onlineSavingsMsg'
Learn More

Loganin, MedChemExpress

Product Code. 16454847
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16454847

Brand: Medchem Express HYN0512

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Loganin, a major iridoid glycoside obtained from Corni fructus, has been shown to have anti-inflammatory and anti-shock effects.

Loganin, a major iridoid glycoside obtained from Corni fructus, has been shown to have anti-inflammatory and anti-shock effects.

Specifications

Chemical Name or Material Loganin
CAS 18524-94-2
Color White
Purity Grade Notes Research
Molecular Formula C17H26O10
Quantity 50 mg
Synonym Loganoside
Solubility Information DMSO : 50 mg/mL (128.08 mM; ultrasonic and warming and heat to 60°C)
SMILES O[C@H]([C@@H](O)[C@@H]1O)[C@](O[C@@H]1CO)([H])O[C@H]2[C@@]([C@H]3C)([H])[C@@](C[C@@H]3O)([H])C(C(OC)=O)=CO2
Molecular Weight (g/mol) 390.38
Formula Weight 390.38
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.