missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lometrexol Hydrate, TRC
Specifications
Specifications
| Chemical Name or Material | Lometrexol Hydrate |
| Synonym | N-[4-[2-[(6R)-2-Amino-3,4,5,6,7,8-hexahydro-4-oxopyrido[2,3-d]pyrimidin-6-yl]ethyl]benzoyl]-L-glutamic Acid Hydrate∣LY 264618 Hydrate∣ |
| Quantity | 5 mg |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H25 N5 O6 . x[H2 O] |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?