missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Lonafarnib
GREENER_CHOICE

Product Code. 18748236
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18748236 10 mg 10mg
18799046 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18748236 Supplier Tocris Bioscience™ Supplier No. 6265/10
  • ONLINE EXCLUSIVE PRICE
    222.00 €
    210.00 € / 10mg
    Save 12.00 €
    5% Off
 Restricted Item
Estimated Shipment: 23-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent farnesyl transferase inhibitor

Lonafarnib is a potent farnesyl transferase inhibitor (IC50 = 1.9 nM). Inhibits farnesylation of RAS. Also inhibits Pgp transport (IC50 < 3 μM) and increases potency and anticancer activity when used in conjunction with cytotoxic Pgp substrates. Inhibits neovascularization by affecting cell motility. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[2-[4-[(11R)-3,10-Dibromo-8-chloro-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-yl]-1-piperidinyl]-2-oxoethyl]-1-piperidinecarboxamide
CAS 193275-84-2
Assay Percent Range >98%
Quantity 10 mg
Molecular Formula C27H31Br2ClN4O2
Formula Weight Observed MW: 638.82
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.