missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Lonafarnib
GREENER_CHOICE

Product Code. p-7114679
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18748236

Brand: Tocris Bioscience™ 6265/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent farnesyl transferase inhibitor

Lonafarnib is a potent farnesyl transferase inhibitor (IC50 = 1.9 nM). Inhibits farnesylation of RAS. Also inhibits Pgp transport (IC50 < 3 μM) and increases potency and anticancer activity when used in conjunction with cytotoxic Pgp substrates. Inhibits neovascularization by affecting cell motility. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[2-[4-[(11R)-3,10-Dibromo-8-chloro-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-yl]-1-piperidinyl]-2-oxoethyl]-1-piperidinecarboxamide
CAS 193275-84-2
Assay Percent Range >98%
Quantity 10 mg
Molecular Formula C27H31Br2ClN4O2
Formula Weight Observed MW: 638.82
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.