missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ Long Life Instrument Syringes

Long life syringes were developed for extreme conditions that include harsh chemicals as well as abrasive particulate matter.

Brand:  Hamilton™ 8300-35

Code : NEW

Additional Details : Weight : 0.06100kg

 View more versions of this product

Product Code. 10748965

  • 195.00 € / Each
Estimated Shipment: 04-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Plunger tip made from UHMWPE (Ultra High Molecular Weight Poly Ethylene) which is harder than PTFE to better resist scratching
  • Plunger rod made from 316 stainless steel which offers exceptional chemical resistance
Specifications

Specifications

Borosilicate Glass, PTFE, UHMWPE
Instrument Syringe
1 mL
13.5 mm
1/Ea.
Luer Lock
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers