missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ Long Life Instrument Syringes

Long life syringes were developed for extreme conditions that include harsh chemicals as well as abrasive particulate matter.

Brand:  Hamilton™ 8300-25

Code : A0

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 11580232

  • 188.00 € / Each
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Plunger tip made from UHMWPE (Ultra High Molecular Weight Poly Ethylene) which is harder than PTFE to better resist scratching
  • Plunger rod made from 316 stainless steel which offers exceptional chemical resistance
Specifications

Specifications

Not Autoclavable
7.7 mm
Gastight Syringe
115°C
10°C
250 μL
Luer Lock
2.3 mm
For corrosive liquids and gases
500 psi
1725
1/Ea.
ML500 Series Diluters and Dispensers
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers