missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ BioWhittaker™ CHO (Chinese Hamster Ovary) Serum-Free Culture Media: ProCHO4

Product Code. 11675140
Change view
Click to view available options
With Additives:
None
Without Additives:
No Phenol Red, No 0.1% Pluronic F-68, No Glutamine, No Hypoxanthine, No Thymidine
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. With Additives Without Additives Unit Size
11675140 None No Phenol Red, No 0.1% Pluronic F-68, No Glutamine, No Hypoxanthine, No Thymidine 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11675140 Supplier Lonza™ Supplier No. 04919Q
  • ONLINE EXCLUSIVE PRICE
    226.00 €
    188.02 € / 1 L
    Save 37.98 €
    16% Off
View all promotions
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For optimized expression of protein in CHO cells

  • ProCHO-4 is designed for the concurrent transition of adherent CHO cells to serum-free and suspension culture; supports faster doubling times

†Store as detailed on label.

TRUSTED_SUSTAINABILITY

Specifications

Cell Line CHO
Form Liquid
With Additives None
Without Additives No Phenol Red, No 0.1% Pluronic F-68, No Glutamine, No Hypoxanthine, No Thymidine
Quantity 1 L
Content And Storage 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.