missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ DNA Ladders

Product Code. 11590777
Change view
Click to view available options
Quantity:
150 μL
160 μL
300 μL
No. of Bands:
10
16
25
30
Concentration:
100 ng/μL
200 ng/μL
Increments:
100bp
20bp
500bp
Unit Size:
100 reactions
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity No. of Bands Concentration Increments unitSize
11590777 300 μL 16 200 ng/μL 500bp 100 reactions
11580777 150 μL 30 200 ng/μL 100bp 100 reactions
10558125 160 μL 10 100 ng/μL 100bp Each
11540787 150 μL 25 200 ng/μL 20bp Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11590777 Supplier Lonza™ Supplier No. 50323

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Each band has 50% GC content for precise sizing and sharp resolution

TRUSTED_SUSTAINABILITY

Specifications

Concentration 200 ng/μL
Form Liquid
Formulation Supplied at various concentrations in TE Buffer and each includes a vial of 6X Sample Loading Buffer 1mL total volume 15% w/v Ficoll 400, 0.06% w/v Bromophenol Blue, 0.06% w/v Xylene cyanol FF, 30mM EDTA
For Use With (Application) Size determination of DNA samples
Increments 500bp
No. of Bands 16
Quantity 300 μL
Storage Requirements 2° to 8°C
Product Type DNA Molecular Weight Marker
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.