missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ FlashGel™ System DNA QuantLadder
Description
- 250μL
- Ready to load
- 50 applications
- No. of Bands: Five
- Sizes (DNA Amount): 100bp (3ng)/ 250bp (7.5ng)/ 400bp (15ng)/ 800bp (21ng)/ 1500bp (30ng)
- Designed to optimize performance of the FlashGel System (Cat. No. BMA57067)
- For research or manufacturing use only
- Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
Specifications
Specifications
| Form | Liquid |
| For Use With (Application) | Size and quantity estimation |
| Quantity | 250 μL, 50 Applications |
| Storage Requirements | 4°C |
| Product Type | DNA Molecular Weight Marker |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?