missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ Formaldehyde Sample Buffer
Description
- For denaturation of RNA samples for RNA electrophoresis
- Ready to use
- Contains formaldehyde, formamide, and tracking dyes in MOPS buffer
Specifications
Specifications
| Buffer | Sample Buffer |
| Form | Liquid |
| Formulation | Contains Formaldehyde, Formamide and Tracking dyes in MOPS Buffer |
| For Use With (Application) | Denaturation of RNA samples for RNA electrophoresis when samples are to be recovered from Agarose gels |
| Quantity | 5 x 1 mL |
| Storage Requirements | −20°C |
| Product Type | RNA Sample Loading Buffer-Formaldehyde |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?