missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ Reliant™ Precast RNA Gels

Product Code. 11537825
Change view
Click to view available options
Wells:
20-well
8-well
Unit Size:
Each
Pack of 20
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wells unitSize
11537825 8-well Pack of 20
11577835 20-well Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11537825 Supplier Lonza™ Supplier No. 54922

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Formulated for separation of RNA of sizes from 0.25 to at least 10kb. Reliant™ Precast RNA Gels are ready-to-use RNase free agarose gels suitable for a variety of RNA applications including Northern blotting and analysis of RNA transcripts.

  • Ready to use
  • RNase free
  • Free of denaturants, eliminating problems associated with staining gels containing formaldehyde
  • Suitable for a variety of applications, including Northern blotting and analysis of RNA transcripts
  • Compatible with most common mini chambers
  • Available with a separate container of glyoxal buffer
TRUSTED_SUSTAINABILITY

Specifications

Buffer MOPS
Concentration 1.25%
Format Precast Gel
Wells 8-well
Dnase Free Yes
Formulation 1.25% SeaKem™ Gold Agarose, 1X MOPS
For Use With (Application) Northern blotting and analysis of RNA transcripts
Material Agarose
Quantity 20 Gels
RNase Free Yes
Storage Requirements 18° to 26°C
Thickness (English) <0.19 in.
Thickness (Metric) <0.5 cm
Volume Per Well <15 μL
Length (English) Gel 3.7 in.
Length (Metric) Gel 9.5 cm
Width (English) Gel 2.37 in.
Width (Metric) Gel 6 cm
Product Type Agarose Gel
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.