missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ RNA Markers

Product Code. 11577745
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11577745 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11577745 Supplier Lonza™ Supplier No. 50575

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For size determination of RNA fragments

  • Consist of RNA transcripts in sizes of 0.5, 1, 1.5, 2, 2.5, 3, 4, 5, 6, and 9kb
  • Can be denatured with standard procedures
  • Can be visualized on Northern blots with labeled lambda sequence
  • Can be detected with ethidium bromide or nucleic acid stains
  • Store at -20°C for up to six months or at -80°C for up to 24 months

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 μg/mL
For Use With (Application) Size determination of RNA fragments
No. of Bands 10
Quantity 50 μg
Storage Requirements −20°C
Product Type RNA Molecular Weight Marker
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.