missing translation for 'onlineSavingsMsg'
Learn More

Loratadine, 98+%, Thermo Scientific™

A nonsedating-type histamine H1-receptor antagonist

88.30 € - 501.00 €

Chemical Identifiers

CAS 79794-75-5
Molecular Formula C22H23ClN2O2
Molecular Weight (g/mol) 382.888
MDL Number MFCD00672869
InChI Key JCCNYMKQOSZNPW-UHFFFAOYSA-N
Synonym loratadine, claritin, loratidine, alavert, clarityn, lisino, clarityne, loracert, loradex, bonalerg
PubChem CID 3957
IUPAC Name ethyl 4-(8-chloro-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridin-11-ylidene)piperidine-1-carboxylate
SMILES CCOC(=O)N1CCC(=C2C3=C(CCC4=C2N=CC=C4)C=C(C=C3)Cl)CC1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15492999
SDS View Documents
Thermo Scientific Alfa Aesar
J60190.ME
500mg
88.30 €
500mg
View alternative products
Save Up to 
»
Estimated Shipment: 21-09-2026
Add to basket
 
 Restricted Item
15472999
SDS View Documents
Thermo Scientific Alfa Aesar
J60190.03
162.00 €
1g
View alternative products
Save Up to 
»
Estimated Shipment: 28-09-2026
Add to basket
 
 Restricted Item
15482999
SDS View Documents
Thermo Scientific Alfa Aesar
J60190.06
5g
501.00 €
5g
View alternative products
Save Up to 
»
Estimated Shipment: 28-09-2026
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Loratadine, is used as a peripheral histamine H1 receptor agonist. It is also used as an orally active antiallergic agent. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
It is soluble in DMSO (50 mg/ml), ethanol (77 mg/ml at 25°C), water (<1mg/mL at 25°C), chloroform, and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away fom strong oxidizing agents.
Specifications

Chemical Identifiers

79794-75-5
382.888
JCCNYMKQOSZNPW-UHFFFAOYSA-N
3957
CCOC(=O)N1CCC(=C2C3=C(CCC4=C2N=CC=C4)C=C(C=C3)Cl)CC1
C22H23ClN2O2
MFCD00672869
loratadine, claritin, loratidine, alavert, clarityn, lisino, clarityne, loracert, loradex, bonalerg
ethyl 4-(8-chloro-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridin-11-ylidene)piperidine-1-carboxylate

Specifications

79794-75-5
MFCD00672869
loratadine, claritin, loratidine, alavert, clarityn, lisino, clarityne, loracert, loradex, bonalerg
JCCNYMKQOSZNPW-UHFFFAOYSA-N
ethyl 4-(8-chloro-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridin-11-ylidene)piperidine-1-carboxylate
3957
≥98%
C22H23ClN2O2
14,5578
It is soluble in DMSO (50mg/ml),ethanol (77mg/ml at 25°C),water (<1mg/ml at 25°C),chloroform,and methanol.
CCOC(=O)N1CCC(=C2C3=C(CCC4=C2N=CC=C4)C=C(C=C3)Cl)CC1
382.888
382.88
Loratadine
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : TM6129200

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.