missing translation for 'onlineSavingsMsg'
Learn More

Loratadine, 98+%, Thermo Scientific™

A nonsedating-type histamine H1-receptor antagonist

Brand:  Thermo Scientific Alfa Aesar J60190.06

Quantity 5g

 View more versions of this product

Product Code. 15482999

  • 501.00 € / 5g
 Restricted Item
Estimated Shipment: 28-09-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Loratadine, is used as a peripheral histamine H1 receptor agonist. It is also used as an orally active antiallergic agent. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
It is soluble in DMSO (50 mg/ml), ethanol (77 mg/ml at 25°C), water (<1mg/mL at 25°C), chloroform, and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away fom strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

79794-75-5
382.888
JCCNYMKQOSZNPW-UHFFFAOYSA-N
3957
CCOC(=O)N1CCC(=C2C3=C(CCC4=C2N=CC=C4)C=C(C=C3)Cl)CC1
C22H23ClN2O2
MFCD00672869
loratadine, claritin, loratidine, alavert, clarityn, lisino, clarityne, loracert, loradex, bonalerg
ethyl 4-(8-chloro-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridin-11-ylidene)piperidine-1-carboxylate

Specifications

79794-75-5
MFCD00672869
14,5578
It is soluble in DMSO (50mg/ml),ethanol (77mg/ml at 25°C),water (<1mg/ml at 25°C),chloroform,and methanol.
CCOC(=O)N1CCC(=C2C3=C(CCC4=C2N=CC=C4)C=C(C=C3)Cl)CC1
382.888
382.88
Loratadine
C22H23ClN2O2
5g
loratadine, claritin, loratidine, alavert, clarityn, lisino, clarityne, loracert, loradex, bonalerg
JCCNYMKQOSZNPW-UHFFFAOYSA-N
ethyl 4-(8-chloro-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridin-11-ylidene)piperidine-1-carboxylate
3957
≥98%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.