missing translation for 'onlineSavingsMsg'
Learn More

Loratadine Impurity F, TRC

Product Code. 30137732
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137732 25 mg 25mg
30134579 100 mg 100mg
30012608 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137732 Supplier Toronto Research Chemicals Supplier No. TRCL46958025MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125743-80-8
Molecular Formula C22 H24 Cl F N2 O2
Molecular Weight (g/mol) 402.89
Synonym 1-Piperidinecarboxylic acid, 4-(8-chloro-11-fluoro-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-yl)-, ethyl ester, 5H-Benzo[5,6]cyclohepta[1,2-b]pyridine, 1-piperidinecarboxylic acid deriv., Ethyl 4-(8-chloro-11-fluoro-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-yl)-1-piperidinecarboxylate, Ethyl 4-[(11RS)-8-chloro-11-fluoro-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-yl]piperidine-1-carboxylate
IUPAC Name ethyl 4-(8-chloro-11-fluoro-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridin-11-yl)piperidine-1-carboxylate
SMILES CCOC(=O)N1CCC(CC1)C2(F)c3ccc(Cl)cc3CCc4cccnc24

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C22H24ClFN2O2/c1-2-28-21(27)26-12-9-17(10-13-26)22(24)19-8-7-18(23)14-16(19)6-5-15-4-3-11-25-20(15)22/h3-4,7-8,11,14,17H,2,5-6,9-10,12-13H2,1H3
Formula Weight 402.151
Percent Purity >95%
Chemical Name or Material ethyl 4-[(11RS)-8-chloro-11-fluoro-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-yl]piperidine-1-carboxylate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.