missing translation for 'onlineSavingsMsg'
Learn More

Loratadine N-Oxide, TRC

Product Code. 30482976
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30482976 50 mg 50mg
30467341 25 mg 25mg
30476640 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30482976 Supplier Toronto Research Chemicals Supplier No. TRCL46959550MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 165739-62-8
Molecular Formula C22 H23 Cl N2 O3
Molecular Weight (g/mol) 398.88
Synonym 1-Piperidinecarboxylic acid, 4-(8-chloro-5,6-dihydro-1-oxido-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)-, ethyl ester, 1-Piperidinecarboxylic acid, 4-(8-chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)-, ethyl ester, N-oxide, Sch 38554, Loratadine N-Oxide
IUPAC Name ethyl 4-(8-chloro-1-oxido-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridin-1-ium-11-ylidene)piperidine-1-carboxylate
SMILES CCOC(=O)N1CCC(=C2c3ccc(Cl)cc3CCc4ccc[n+]([O-])c24)CC1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C22H23ClN2O3/c1-2-28-22(26)24-12-9-15(10-13-24)20-19-8-7-18(23)14-17(19)6-5-16-4-3-11-25(27)21(16)20/h3-4,7-8,11,14H,2,5-6,9-10,12-13H2,1H3
Formula Weight 398.1397
Chemical Name or Material Loratadine N-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.