missing translation for 'onlineSavingsMsg'
Learn More

Lorcaserin Hydrochloride (>50% ee), TRC

Product Code. 30136009
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136009 25 mg 25mg
30136874 50 mg 50mg
30009026 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136009 Supplier Toronto Research Chemicals Supplier No. TRCL46989025MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 846589-98-8
Molecular Formula C11 H14 Cl N . Cl H
Molecular Weight (g/mol) 232.15
Synonym 1H-3-Benzazepine, 8-chloro-2,3,4,5-tetrahydro-1-methyl-, hydrochloride (1:1), (1R)-, 1H-3-Benzazepine, 8-chloro-2,3,4,5-tetrahydro-1-methyl-, hydrochloride, (1R)- (9CI), (R)-8-Chloro-1-methyl-2,3,4,5-tetrahydro-1H-3-benzazepine hydrochloride, (R)-8-Chloro-1-methyl-2,3,4,5-tetrahydro-1H-benzo[d]azepine hydrochloride, APD 356, Lorcaserin hydrochloride
IUPAC Name (5R)-7-chloro-5-methyl-2,3,4,5-tetrahydro-1H-3-benzazepine;hydrochloride
SMILES Cl.C[C@H]1CNCCc2ccc(Cl)cc12

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C11H14ClN.ClH/c1-8-7-13-5-4-9-2-3-10(12)6-11(8)9;/h2-3,6,8,13H,4-5,7H2,1H3;1H/t8-;/m0./s1
Formula Weight 231.0582
Percent Purity >95%
Chemical Name or Material Lorcaserin Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.